| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
NM_001163239.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
NM_001163239.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
NM_001163241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
NM_001163241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
NM_020282.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
NM_020282.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002293.mm.1 |
TC1300000383.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
NM_001163241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
NM_001163241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
NM_001163242.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
NM_001163242.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
NM_020282.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
NM_020282.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002299.mm.1 |
TC1300000383.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
NM_001163239.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
NM_001163239.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
NM_001163241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
NM_001163241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
NM_020282.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
NM_020282.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300002292.mm.1 |
TC1300000383.mm.1 |
MTA TC |