Search
Search
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198977.1 | NP_001185906.1 | 8 - 9 | 1197 | 78 | |
| NM_011518.2 | NP_035648.2 | 8 - 9 | 1201 | 78 | ||
| GenBank mRNA | AK049207.1 | - | 8 - 9 | 1226 | 78 | |
| AK136363.1 | - | 8 - 9 | 1200 | 78 | ||
| AK143564.1 | - | 8 - 9 | 1230 | 78 | ||
| BC065121.1 | - | 8 - 9 | 1133 | 30533300 | 78 | |
| U25685.1 | - | 7 - 8 | 985 | 78 | ||
| U36776.1 | - | 8 - 9 | 1204 | 78 | ||
Set Membership: |
|