Search
Search
| Gene Symbol: Pygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153781.1 | NP_722476.1 | 8 - 9 | 1066 | 90 | |
| GenBank mRNA | AK032769.1 | - | 8 - 9 | 1079 | 90 | |
| AK035673.1 | - | 8 - 9 | 796 | 90 | ||
| AK134667.1 | - | 8 - 9 | 1076 | 90 | ||
| AK142716.1 | - | 8 - 9 | 1076 | 90 | ||
| AK147763.1 | - | 8 - 9 | 1098 | 90 | ||
| AK168381.1 | - | 8 - 9 | 1077 | 90 | ||
| AK169052.1 | - | 8 - 9 | 1101 | 90 | ||
| AK169631.1 | - | 8 - 9 | 1080 | 90 | ||
| BC031457.1 | - | 8 - 9 | 1074 | 90 | ||
| BC032209.1 | - | 2 - 3 | 221 | 90 | ||
| BC035283.1 | - | 8 - 9 | 1066 | 4947455 | 90 | |
Set Membership: |
|