Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333146_g1
          See other LCE3B GE Assays ›
          Gene Symbol:
          Lce3b
          Gene Name:
          late cornified envelope 3B
          Gene Aliases:
          2310007F04Rik, Sprrl6A
          Chromosome Location:
          Chr.3: 92932979 - 92934096 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333146_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lce3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025501.3 NP_079777.1 1 - 2 44 64
          GenBank mRNA AK009001.1 - 1 - 2 45 64
          AK009201.1 - 1 - 2 46 64
          BC115788.1 - 1 - 2 43 40105038 64
          BC115817.1 - 1 - 2 37 40105036 64

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK009001.1, AK009201.1, BC115788.1, BC115817.1, NM_025501.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lce3b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101930 AK009001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101930 AK009201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101930 BC059084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101930 BC115788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101930 BC115817.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478050 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101930 AK009001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101930 AK009201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101930 BC059084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101930 BC115788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101930 BC115817.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22333743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22333733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22333737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22333744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22333735 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04990263 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101930 AK009001.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101930 AK009201.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101930 BC059084.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101930 BC115788.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101930 BC115817.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03111782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22333674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02672908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03111765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03107821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02672902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03111782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22333742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03111767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493904 - -
          Affymetrix HT MG-430 PM Array Plate 1429641_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493896 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493903 - -
          Mouse Genome 430 2.0 Array 1429641_PM_x_at - -
          Mouse Genome 430 2.0 Array 1429641_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66344_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66344_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006431.mm.1 NM_025501.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006431.mm.1 NM_025501.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006431.mm.1 TC0300000834.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006429.mm.1 NM_025501.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006429.mm.1 NM_025501.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006429.mm.1 TC0300000834.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006430.mm.1 NM_025501.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006430.mm.1 NM_025501.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006430.mm.1 TC0300000834.mm.1 MTA TC
          Mouse Expression Set 430 1429641_x_at - -
          Mouse Expression Set 430 1429641_PM_x_at - -
          Murine Genome U74v2 Set 165963_f_at - -
          Murine Genome U74v2 Set 165963_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429641_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1429641_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 434609 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170152 - -
          Illumina mouse-6 v1.0 expression beadchip 4810400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621158 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621158 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619519 - -
          mouse-6 v1.1 (Illumina) scl0066344.1_3-S - -
          mouse-6 v1.1 (Illumina) scl066344.2_272-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810400 - -
          Illumina Sentrix mouse-6.v1 bead array 3170152 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066344.2_272-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066344.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2941749 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2619519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621158 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2941749 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619519 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621158 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline