Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333323_g1
          See other HSPA5 GE Assays ›
          Gene Symbol:
          Hspa5
          Gene Name:
          heat shock protein 5
          Gene Aliases:
          AL022860, AU019543, Bip, D2Wsu141e, D2Wsu17e, Grp78, Hsce70, SEZ-7, Sez7, baffled, mBiP
          Chromosome Location:
          Chr.2: 34772090 - 34776529 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333323_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hspa5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163434.1 NP_001156906.1 7 - 8 1387 92
          NM_022310.3 NP_071705.3 6 - 7 1469 92
          GenBank mRNA AJ002387.1 - 6 - 7 1248 92
          AK004578.1 - 6 - 7 1464 92
          AK076079.1 - 7 - 8 1383 92
          AK077463.1 - 7 - 8 1388 92
          AK144650.1 - 7 - 8 1384 92
          AK144742.1 - 7 - 8 1385 92
          AK146647.1 - 7 - 8 1386 92
          AK148539.1 - 7 - 8 1386 92
          AK149447.1 - 7 - 8 1386 92
          AK151244.1 - 6 - 7 1336 92
          AK151647.1 - 7 - 8 1385 92
          AK151806.1 - 7 - 8 1386 92
          AK152020.1 - 7 - 8 1383 92
          AK152521.1 - 6 - 7 1466 92
          AK152958.1 - 7 - 8 1386 92
          AK159719.1 - 7 - 8 92
          AK160592.1 - 2 - 3 387 92
          AK160821.1 - 1386 92
          AK166207.1 - 7 - 8 1384 92
          AK166739.1 - 7 - 8 1383 92
          AK167011.1 - 7 - 8 1383 92
          AK168012.1 - 7 - 8 1387 92
          AK168230.1 - 7 - 8 1381 92
          AK169034.1 - 7 - 8 1387 92
          AK196010.1 - 1 - 2 41 92
          AK204137.1 - 1 - 2 41 92
          AK206130.1 - 1 - 2 41 92
          AK207319.1 - 1 - 2 41 92
          AK207523.1 - 1 - 2 41 92
          AK207843.1 - 1 - 2 41 92
          AK216390.1 - 1 - 2 41 92
          AK219238.1 - 1 - 2 41 92
          BC005785.1 - 5 - 6 1103 92
          BC050927.1 - 6 - 7 1446 6415084 92
          D78645.1 - 6 - 7 1329 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AY351588.1 - Chr. - - [-] on Build 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000620.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hspa5

          Gene Ontology Categories:

          Function(s) Process(es)

          ER overload response
          activation of signaling protein activity involved in unfolded protein response
          cerebellum structural organization
          cerebellar Purkinje cell layer development
          positive regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of protein ubiquitination
          response to endoplasmic reticulum stress
          maintenance of protein localization in endoplasmic reticulum
          positive regulation of embryonic development
          cellular response to glucose starvation
          negative regulation of apoptotic process
          proteolysis involved in cellular protein catabolic process
          cellular response to interleukin-4
          toxin transport
          nucleotide binding
          glycoprotein binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          ribosome binding
          unfolded protein binding
          misfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129650 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129648 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129649 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129647 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076772 BC050927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076772 D78645.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076772 M30779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076772 AK168230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076772 AK169034.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10560 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076772 BC050927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076772 D78645.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076772 M30779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076772 AK168230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076772 AK169034.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395098 BC050927.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395098 D78645.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395098 M19351.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395098 M30779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395098 AK218616.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21129660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00342422 - -
          Whole Mouse Genome G4122A A_51_P395098 BC050927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395098 D78645.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395098 M19351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395098 M30779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395098 AK218616.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116008 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00342422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02072158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02072144 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02072145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01311713 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02072142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02072156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129650 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129648 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21129649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21129631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02048291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02048292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02048296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01524855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01263380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01442809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21129651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471592 - -
          Affymetrix HT MG-430 PM Array Plate 1416064_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1447824_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1427464_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10471586 - -
          Mouse Genome 430 2.0 Array 1427464_s_at - -
          Mouse Genome 430 2.0 Array 1447824_PM_x_at - -
          Mouse Genome 430 2.0 Array 1416064_PM_a_at - -
          Mouse Genome 430 2.0 Array 1427464_PM_s_at - -
          Mouse Genome 430 2.0 Array 1447824_x_at - -
          Mouse Genome 430 2.0 Array 1416064_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14828_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14828_at - -
          Affymetrix Mouse Clariom S Array PSR0200005688.mm.1 NM_001163434.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005688.mm.1 NM_001163434.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005688.mm.1 NM_022310.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005688.mm.1 NM_022310.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005688.mm.1 TC0200000620.mm.1 MTA TC
          Mouse Expression Set 430 1427464_s_at - -
          Mouse Expression Set 430 1416064_PM_a_at - -
          Mouse Expression Set 430 1416064_a_at - -
          Mouse Expression Set 430 1427464_PM_s_at - -
          Mouse Expression Set 430 1447824_x_at - -
          Mouse Expression Set 430 1447824_PM_x_at - -
          Murine Genome U74v2 Set 101955_PM_at - -
          Murine Genome U74v2 Set 101955_at - -
          Mouse Genome 430A 2.0 Array 1416064_a_at - -
          Mouse Genome 430A 2.0 Array 1427464_s_at - -
          Mouse Genome 430A 2.0 Array 1427464_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1416064_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416064_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427464_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416064_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427464_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1447824_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447824_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5130671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255561 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255561 - -
          mouse-6 v1.1 (Illumina) scl014828.8_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014828.8_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255561 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255561 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl014828.8_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline