Search
Search
| Gene Symbol: Ola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025942.2 | NP_080218.1 | 10 - 11 | 1255 | 145 | |
| GenBank mRNA | AK010817.1 | - | 10 - 11 | 1171 | 145 | |
| AK013055.1 | - | 10 - 11 | 1256 | 145 | ||
| AK159873.1 | - | 10 - 11 | 1172 | 145 | ||
| AK167458.1 | - | 10 - 11 | 1171 | 145 | ||
| AK170982.1 | - | 10 - 11 | 1186 | 145 | ||
| BC011318.1 | - | 10 - 11 | 1155 | 3985271 | 145 | |
| BC029207.1 | - | 10 - 11 | 1148 | 4009561 | 145 | |
Set Membership: |
|