Search
Search

Dync1i2| Gene Symbol: Dync1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198872.1 | NP_001185801.1 | 9 - 10 | 813 | 182 | |
| NM_001198873.1 | NP_001185802.1 | 9 - 10 | 798 | 182 | ||
| NM_001198874.1 | NP_001185803.1 | 7 - 8 | 744 | 182 | ||
| NM_001198875.1 | NP_001185804.1 | 7 - 8 | 702 | 182 | ||
| NM_001198876.1 | NP_001185805.1 | 6 - 7 | 684 | 182 | ||
| NM_001198877.1 | NP_001185806.1 | 9 - 10 | 802 | 182 | ||
| NM_001198878.1 | NP_001185807.1 | 9 - 10 | 787 | 182 | ||
| NM_010064.4 | NP_034194.1 | 6 - 7 | 673 | 182 | ||
| GenBank mRNA | AF063231.1 | - | 6 - 7 | 612 | 182 | |
| AK088015.1 | - | 6 - 7 | 644 | 182 | ||
| AK141837.1 | - | 7 - 8 | 712 | 182 | ||
| AK150173.1 | - | 6 - 7 | 634 | 182 | ||
| AK161831.1 | - | 2 - 3 | 1334 | 182 | ||
| AK164322.1 | - | 7 - 8 | 668 | 182 | ||
| AK164437.1 | - | 6 - 7 | 608 | 182 | ||
| AK168730.1 | - | 6 - 7 | 648 | 182 | ||
| BC139374.1 | - | 6 - 7 | 568 | 8862396 | 182 | |
| BC139375.1 | - | 6 - 7 | 568 | 8862397 | 182 | |
| GU992208.1 | - | 9 - 10 | 704 | 182 | ||
| GU992209.1 | - | 9 - 10 | 689 | 182 | ||
| GU992210.1 | - | 7 - 8 | 593 | 182 | ||
| GU992211.1 | - | 9 - 10 | 742 | 182 | ||
| GU992212.1 | - | 9 - 10 | 727 | 182 | ||
Set Membership: |
|