Search
Search
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244198.1 | NP_001231127.1 | 6 - 7 | 689 | 97 | |
| NM_001244200.1 | NP_001231129.1 | 7 - 8 | 901 | 97 | ||
| NM_001244201.1 | NP_001231130.1 | 5 - 6 | 647 | 97 | ||
| NM_001244202.1 | NP_001231131.1 | 5 - 6 | 1203 | 97 | ||
| NM_013627.5 | NP_038655.1 | 6 - 7 | 1245 | 97 | ||
| GenBank mRNA | AJ292077.1 | - | 5 - 6 | 506 | 97 | |
| AJ307468.1 | - | 5 - 6 | 513 | 97 | ||
| AK028059.1 | - | 5 - 6 | 648 | 97 | ||
| AK032616.1 | - | 6 - 7 | 690 | 97 | ||
| AK045805.1 | - | 5 - 6 | 1204 | 97 | ||
| BC011272.1 | - | 6 - 7 | 933 | 4008490 | 97 | |
| BC036957.1 | - | 5 - 6 | 643 | 4504106 | 97 | |
| M77842.1 | - | 1 - 2 | 232 | 97 | ||
| X63963.1 | - | 6 - 7 | 566 | 97 | ||
Set Membership: |
|