Search
Search

Dcaf17| Gene Symbol: Dcaf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165980.1 | NP_001159452.1 | 12 - 13 | 1513 | 72 | |
| NM_001165981.1 | NP_001159453.1 | 12 - 13 | 1513 | 72 | ||
| NM_198005.2 | NP_932122.2 | 12 - 13 | 1513 | 72 | ||
| GenBank mRNA | AK014717.1 | - | 12 - 13 | 1477 | 72 | |
| AK081839.1 | - | 12 - 13 | 1512 | 72 | ||
| AK160679.1 | - | 12 - 13 | 1480 | 72 | ||
| BB772704.1 | - | 2 - 3 | 118 | 72 | ||
| BC031546.1 | - | 12 - 13 | 1405 | 3669736 | 72 | |
Set Membership: |
|