Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01334345_m1
          See other DCAF17 GE Assays ›
          Gene Symbol:
          Dcaf17
          Gene Name:
          DDB1 and CUL4 associated factor 17
          Gene Aliases:
          2810055O12Rik, 4833418A01Rik, A030004A10Rik, A930009G19Rik, AI448937
          Chromosome Location:
          Chr.2: 71055742 - 71114514 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01334345_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dcaf17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001165980.1 NP_001159452.1 6 - 7 867 76
          NM_001165981.1 NP_001159453.1 6 - 7 867 76
          NM_001165982.1 NP_001159454.1 6 - 7 867 76
          NM_198005.2 NP_932122.2 6 - 7 867 76
          GenBank mRNA AK014717.1 - 6 - 7 831 76
          AK032671.1 - 6 - 7 829 76
          AK079462.1 - 6 - 7 661 76
          AK081839.1 - 6 - 7 866 76
          AK160679.1 - 6 - 7 834 76
          BC031546.1 - 6 - 7 759 3669736 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001030.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dcaf17

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_001165980.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_001165980.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_001165981.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_001165981.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_001165982.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_001165982.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_198005.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 NM_198005.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008266.mm.1 TC0200001030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_001165980.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_001165980.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_001165981.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_001165981.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_001165982.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_001165982.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_198005.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 NM_198005.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008265.mm.1 TC0200001030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008270.mm.1 NM_001165982.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008270.mm.1 NM_001165982.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008270.mm.1 TC0200001030.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline