Search
Search

Cage1| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027724.2 | NP_082000.2 | 5 - 6 | 2043 | 92 | |
| GenBank mRNA | AK132733.1 | - | 1 - 2 | 591 | 92 | |
| AY695827.1 | - | 4 - 5 | 1778 | 92 | ||
Set Membership: |
|