Search
Search

Tasp1| Gene Symbol: Tasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159640.2 | NP_001153112.1 | 1 - 2 | 48 | 70 | |
| NM_001159641.2 | NP_001153113.1 | 1 - 2 | 48 | 70 | ||
| NM_001289611.1 | NP_001276540.1 | 1 - 2 | 48 | 70 | ||
| NR_110351.1 | 1 - 2 | 48 | 70 | |||
| NR_110352.1 | 1 - 2 | 48 | 70 | |||
| GenBank mRNA | AK015624.1 | - | 1 - 2 | 48 | 70 | |
Set Membership: |
|