Search
Search
| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172666.3 | NP_766254.2 | 2 - 3 | 424 | 73 | |
| GenBank mRNA | AK031049.1 | - | 2 - 3 | 426 | 73 | |
| AK031154.1 | - | 2 - 3 | 265 | 73 | ||
| AK042745.1 | - | 2 - 3 | 264 | 73 | ||
| AK053686.1 | - | 2 - 3 | 413 | 73 | ||
| AK079447.1 | - | 1 - 2 | 104 | 73 | ||
| BC063086.1 | - | 2 - 3 | 409 | 6332574 | 73 | |
| BY713473.1 | - | 2 - 3 | 424 | 73 | ||
Set Membership: |
|