Search
Search

Alkbh3| Gene Symbol: Alkbh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026944.1 | NP_081220.1 | 5 - 6 | 381 | 70 | |
| GenBank mRNA | AK145586.1 | - | 5 - 6 | 767 | 70 | |
| BC018196.1 | - | 5 - 6 | 381 | 3993214 | 70 | |
Set Membership: |
|