Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335362_g1
          See other LILRA5 GE Assays ›
          Gene Symbol:
          Lilra5
          Gene Name:
          leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5
          Gene Aliases:
          E030017K20, EG232801, Gm4878, Lilrc1
          Chromosome Location:
          Chr.7: 4237643 - 4243690 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335362_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lilra5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081239.2 NP_001074708.2 5 - 6 716 76
          GenBank mRNA AK086983.1 - 2 - 3 129 76
          AY998482.1 - 5 - 6 716 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000028.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991560 NM_001081239.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991560 AK086983.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690730 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991560 NM_001081239.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991560 AK086983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364185 NM_001081239.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364185 AK086983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65610 NM_001081239.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P65610 AY998482.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24948222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06598790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06390396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06598787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06390399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06598785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06598783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06011977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06598757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549639 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549635 - -
          Affymetrix Mouse Clariom S Array PSR0700000385.mm.1 NM_001081239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000385.mm.1 NM_001081239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000385.mm.1 TC0700000028.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 692170 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2784514 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2784514 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline