Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335415_g1
          See other THBS1 GE Assays ›
          Gene Symbol:
          Thbs1
          Gene Name:
          thrombospondin 1
          Gene Aliases:
          TSP-1, TSP1, Thbs-1, tbsp1
          Chromosome Location:
          Chr.2: 118111890 - 118127133 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335415_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thbs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011580.3 NP_035710.2 14 - 15 2444 77
          GenBank mRNA AK080686.1 - 14 - 15 2479 77
          AK145202.1 - 14 - 15 2474 77
          AK163092.1 - 14 - 15 2471 77
          BC042422.1 - 14 - 15 2446 4018176 77
          BC050917.1 - 14 - 15 2444 6405710 77
          M87276.1 - 14 - 15 2470 77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK089315.1 - Chr. - - [-] on Build 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001729.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thbs1

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          negative regulation of endothelial cell proliferation
          negative regulation of cell-matrix adhesion
          sprouting angiogenesis
          negative regulation of antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          negative regulation of dendritic cell antigen processing and presentation
          outflow tract morphogenesis
          endocardial cushion development
          growth plate cartilage development
          inflammatory response
          response to unfolded protein
          cell cycle arrest
          cell adhesion
          response to mechanical stimulus
          response to glucose
          positive regulation of endothelial cell migration
          negative regulation of endothelial cell migration
          negative regulation of plasma membrane long-chain fatty acid transport
          negative regulation of nitric oxide mediated signal transduction
          negative regulation of cGMP-mediated signaling
          negative regulation of plasminogen activation
          positive regulation of macrophage chemotaxis
          positive regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          cell migration
          negative regulation of angiogenesis
          peptide cross-linking
          positive regulation of blood coagulation
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          regulation of cGMP metabolic process
          response to magnesium ion
          negative regulation of interleukin-12 production
          response to endoplasmic reticulum stress
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of phosphorylation
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of macrophage activation
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          engulfment of apoptotic cell
          positive regulation of translation
          positive regulation of angiogenesis
          behavioral response to pain
          blood vessel morphogenesis
          positive regulation of chemotaxis
          response to calcium ion
          positive regulation of protein kinase B signaling
          negative regulation of fibrinolysis
          positive regulation of transforming growth factor beta production
          cellular response to nitric oxide
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          negative regulation of endothelial cell chemotaxis
          phosphatidylserine binding
          fibronectin binding
          integrin binding
          calcium ion binding
          protein binding
          heparin binding
          fibroblast growth factor binding
          low-density lipoprotein particle binding
          laminin binding
          transforming growth factor beta binding
          extracellular matrix binding
          fibrinogen binding
          collagen V binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline