Search
Search

Trip11| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028446.1 | NP_082722.1 | 13 - 14 | 5230 | 88 | |
| GenBank mRNA | BC044791.1 | - | 2 - 3 | 342 | 88 | |
| BC158098.1 | - | 13 - 14 | 5326 | 9088179 | 88 | |
Set Membership: |
|