Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336355_g1
          See other SMTNL1 GE Assays ›
          Gene Symbol:
          Smtnl1
          Gene Name:
          smoothelin-like 1
          Gene Aliases:
          1110030K22Rik, CHASM
          Chromosome Location:
          Chr.2: 84811176 - 84826934 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336355_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smtnl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024230.2 NP_077192.1 5 - 6 1131 81
          GenBank mRNA AK003989.1 - 5 - 6 1024 81
          BC002317.1 - 5 - 6 1131 3593616 81

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smtnl1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to activity
          peptidyl-serine phosphorylation
          response to drug
          negative regulation of catalytic activity
          positive regulation of vasoconstriction
          negative regulation of vasodilation
          muscle organ morphogenesis
          protein phosphatase inhibitor activity
          protein binding
          calmodulin binding
          protein phosphatase 1 binding
          myosin phosphatase regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993153 BC002317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993153 NM_024230.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168521 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107381 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993153 BC002317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993153 NM_024230.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394808 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04591501 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168521 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107381 - -
          Whole Mouse Genome G4122A A_51_P359424 BC002317.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128516 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107381 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168521 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02006916 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02006908 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02173324 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02006909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02173326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01803851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02006920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01603302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01462776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01462782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01264586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01859567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02006914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01645126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02006919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01462777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01937088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394753 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484477 - -
          Affymetrix HT MG-430 PM Array Plate 1419145_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10484472 - -
          Mouse Genome 430 2.0 Array 1419145_at - -
          Mouse Genome 430 2.0 Array 1419145_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68678_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68678_at - -
          Mouse Expression Set 430 1419145_at - -
          Mouse Expression Set 430 1419145_PM_at - -
          Murine Genome U74v2 Set 165685_at - -
          Murine Genome U74v2 Set 165685_PM_at - -
          Mouse Genome 430A 2.0 Array 1419145_PM_at - -
          Mouse Genome 430A 2.0 Array 1419145_at - -
          Affymetrix HT Mouse Genome 430A Array 1419145_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419145_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 384673 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060278 - -
          Illumina mouse-6 v1.0 expression beadchip 3060278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774281 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774281 - -
          mouse-6 v1.1 (Illumina) scl19065.8.1_153-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19065.8.1_153-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2774281 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774281 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19065.8.1_153-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline