Search
Search

Calcrl| Gene Symbol: Calcrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018782.2 | NP_061252.2 | 5 - 6 | 571 | 125 | |
| GenBank mRNA | AB015595.1 | - | 5 - 6 | 575 | 125 | |
| AF146525.1 | - | 3 - 4 | 248 | 125 | ||
| AK137210.1 | - | 5 - 6 | 572 | 125 | ||
| BC138537.1 | - | 2 - 3 | 198 | 8861557 | 125 | |
| BC138540.1 | - | 2 - 3 | 198 | 8861560 | 125 | |
Set Membership: |
|