Search
Search

Rab14| Gene Symbol: Rab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026697.3 | NP_080973.1 | 5 - 6 | 717 | 70 | |
| GenBank mRNA | AB232609.1 | - | 4 - 5 | 348 | 70 | |
| AJ968955.1 | - | 5 - 6 | 572 | 70 | ||
| AJ968956.1 | - | 4 - 5 | 518 | 70 | ||
| AJ968957.1 | - | 5 - 6 | 572 | 70 | ||
| AJ968958.1 | - | 4 - 5 | 518 | 70 | ||
| AJ968959.1 | - | 5 - 6 | 572 | 70 | ||
| AJ968960.1 | - | 4 - 5 | 518 | 70 | ||
| AK002704.1 | - | 5 - 6 | 589 | 70 | ||
| AK013410.1 | - | 5 - 6 | 570 | 70 | ||
| AK133963.1 | - | 5 - 6 | 617 | 70 | ||
| AK135751.1 | - | 5 - 6 | 606 | 70 | ||
| AK160859.1 | - | 5 - 6 | 717 | 70 | ||
| AK165812.1 | - | 5 - 6 | 564 | 70 | ||
| AK168551.1 | - | 5 - 6 | 574 | 70 | ||
| AK168867.1 | - | 5 - 6 | 586 | 70 | ||
| AK169396.1 | - | 5 - 6 | 588 | 70 | ||
| AK186545.1 | - | 2 - 3 | 125 | 70 | ||
| BC009085.1 | - | 5 - 6 | 400 | 2649852 | 70 | |
| BC025139.1 | - | 5 - 6 | 588 | 3980228 | 70 | |
| BC056648.1 | - | 5 - 6 | 590 | 5034519 | 70 | |
Set Membership: |
|