Search
Search
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 18 - 19 | 2421 | 107 | |
| GenBank mRNA | M35525.1 | - | 18 - 19 | 2376 | 107 | |
| M35526.1 | - | 18 - 19 | 2374 | 107 | ||
Set Membership: |
|