Search
Search

9030617O03Rik| Gene Symbol: 9030617O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145448.4 | NP_663423.2 | 7 - 8 | 1139 | 94 | |
| GenBank mRNA | AK033551.1 | - | 7 - 8 | 1168 | 94 | |
| AK038436.1 | - | 7 - 8 | 1141 | 94 | ||
| AK081256.1 | - | 1176 | 94 | |||
| AK152622.1 | - | 7 - 8 | 1168 | 94 | ||
| BC021385.1 | - | 8 - 9 | 1124 | 5064508 | 94 | |
| BC066161.1 | - | 8 - 9 | 1213 | 30093441 | 94 | |
Set Membership: |
|