Search
Search

Spata7| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289572.1 | NP_001276501.1 | 7 - 8 | 1069 | 117 | |
| NM_001289573.1 | NP_001276502.1 | 6 - 7 | 664 | 117 | ||
| NM_001289574.1 | NP_001276503.1 | 8 - 9 | 1231 | 117 | ||
| NM_178914.4 | NP_849245.2 | 8 - 9 | 1165 | 117 | ||
| GenBank mRNA | AK036359.1 | - | 8 - 9 | 1149 | 117 | |
| AK077500.1 | - | 8 - 9 | 1076 | 117 | ||
| AK153698.1 | - | 6 - 7 | 640 | 117 | ||
| AK161308.1 | - | 7 - 8 | 1015 | 117 | ||
| BC046960.1 | - | 8 - 9 | 1123 | 6391450 | 117 | |
| DQ196101.1 | - | 7 - 8 | 895 | 117 | ||
Set Membership: |
|