Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01337129_g1
          See other NAA38 GE Assays ›
          Gene Symbol:
          Naa38
          Gene Name:
          N(alpha)-acetyltransferase 38, NatC auxiliary subunit
          Gene Aliases:
          1500034E06Rik, Lsmd1
          Chromosome Location:
          Chr.11: 69395791 - 69396671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01337129_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Naa38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030083.2 NP_084359.1 2 - 3 365 95
          GenBank mRNA AK018782.1 - 2 - 3 366 95
          BC038659.1 - 2 - 3 319 95
          BC048499.1 - 2 - 3 371 6583055 95
          BC062267.1 - 2 - 3 335 1532489 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000912.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Naa38

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of apoptotic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894826 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894825 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 BC048499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 BC062267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 AK018782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 BC038659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 NM_030083.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 BC048499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 BC062267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 AK018782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 BC038659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314323 NM_030083.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491462 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147923 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 BC048499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 BC062267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 AK018782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 BC038659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 NM_030083.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 BC048499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 BC062267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 AK018782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 BC038659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314323 NM_030083.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27894831 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147923 - -
          Whole Mouse Genome G4122A A_51_P314323 BC048499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 BC062267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 AK018782.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 BC038659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 NM_030083.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 BC048499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 BC062267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 AK018782.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 BC038659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314323 NM_030083.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102870 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09694635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09694645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894826 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27894827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27894903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27894829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27894825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27894827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27894824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27894822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27894831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377544 - -
          Affymetrix HT MG-430 PM Array Plate 1428526_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10387368 - -
          Affymetrix Mouse Gene 1.1 ST Array 10377541 - -
          Mouse Genome 430 2.0 Array 1428526_at - -
          Mouse Genome 430 2.0 Array 1428526_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78304_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78304_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100007728.mm.1 TC1100000912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007730.mm.1 NM_030083.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007730.mm.1 NM_030083.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007730.mm.1 TC1100000912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007731.mm.1 NM_030083.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007731.mm.1 NM_030083.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007731.mm.1 TC1100000912.mm.1 MTA TC
          Mouse Expression Set 430 1428526_at - -
          Mouse Expression Set 430 1428526_PM_at - -
          Murine Genome U74v2 Set 97271_PM_at - -
          Murine Genome U74v2 Set 161860_PM_f_at - -
          Murine Genome U74v2 Set 161860_f_at - -
          Murine Genome U74v2 Set 97271_at - -
          Murine Genome U74v2 Set 171468_PM_i_at - -
          Murine Genome U74v2 Set 171468_i_at - -
          Mouse Genome 430A 2.0 Array 1428526_at - -
          Mouse Genome 430A 2.0 Array 1428526_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428526_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428526_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309126 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257994 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257994 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline