Search
Search
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ223765.1 | - | 3 - 4 | 373 | 96 | |
| AK172251.1 | - | 2 - 3 | 417 | 96 | ||
| BC042788.1 | - | 3 - 4 | 354 | 96 | ||
| BC115364.2 | - | 3 - 4 | 385 | 40045898 | 96 | |
Set Membership: |
|