Search
Search
| Gene Symbol: Psd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177611.3 | NP_808279.1 | 13 - 14 | 2478 | 77 | |
| GenBank mRNA | AK043669.1 | - | 14 - 15 | 2770 | 77 | |
| AK148489.1 | - | 3 - 4 | 253 | 77 | ||
| AK150238.1 | - | 13 - 14 | 2507 | 77 | ||
| AK155249.1 | - | 13 - 14 | 2508 | 77 | ||
| AK163628.1 | - | 9 - 10 | 857 | 77 | ||
| AK169859.1 | - | 13 - 14 | 2507 | 77 | ||
| BC046518.1 | - | 5 - 6 | 405 | 77 | ||
| BC068231.1 | - | 13 - 14 | 2478 | 30361859 | 77 | |
Set Membership: |
|