Search
Search

Tgfbi| Gene Symbol: Tgfbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009369.4 | NP_033395.1 | 13 - 14 | 1846 | 76 | |
| GenBank mRNA | AK084431.1 | - | 13 - 14 | 1864 | 76 | |
| AK135536.1 | - | 13 - 14 | 1891 | 76 | ||
| AK142323.1 | - | 13 - 14 | 1866 | 76 | ||
| AK150449.1 | - | 13 - 14 | 1864 | 76 | ||
| AK150833.1 | - | 13 - 14 | 1864 | 76 | ||
| AK150969.1 | - | 13 - 14 | 1865 | 76 | ||
| AK151680.1 | - | 13 - 14 | 1861 | 76 | ||
| AK152365.1 | - | 13 - 14 | 1865 | 76 | ||
| AK152662.1 | - | 13 - 14 | 1864 | 76 | ||
| AK155572.1 | - | 13 - 14 | 1861 | 76 | ||
| AK155828.1 | - | 13 - 14 | 1861 | 76 | ||
| AK159711.1 | - | 13 - 14 | 1860 | 76 | ||
| AK164373.1 | - | 13 - 14 | 1864 | 76 | ||
| AK170495.1 | - | 13 - 14 | 1861 | 76 | ||
| AK171557.1 | - | 12 - 13 | 1685 | 76 | ||
| BC129900.1 | - | 13 - 14 | 1839 | 40111549 | 76 | |
| BC129901.1 | - | 13 - 14 | 1839 | 40111550 | 76 | |
| L19932.1 | - | 13 - 14 | 1847 | 76 | ||
Set Membership: |
|