Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338573_s1
          See other OLFR1162 GE Assays ›
          Gene Symbol:
          Olfr1162
          Gene Name:
          olfactory receptor 1162
          Gene Aliases:
          MOR174-14
          Chromosome Location:
          Chr.2: 88049678 - 88050622 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338573_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1162
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011835.1 NP_001011835.1 1 894 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1162

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147706 NM_001011835.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147706 NM_001011835.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01543714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01990984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02248090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01990981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01990962 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484713 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484712 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897195 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162441 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162441 - -
          mouse-6 v1.1 (Illumina) GI_58801433-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162441 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline