Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338840_s1
          See other OLFR1238 GE Assays ›
          Gene Symbol:
          Olfr1238
          Gene Name:
          olfactory receptor 1238
          Gene Aliases:
          MOR231-11
          Chromosome Location:
          Chr.2: 89406130 - 89407077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338840_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1238
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146790.1 NP_667001.1 1 885 153
          GenBank mRNA BC128571.1 - 1 887 40135708 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1238

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006688 NM_146790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006688 BC128571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006688 NM_146790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006688 BC128571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124209 NM_146790.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124209 BC128571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124209 NM_146790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124209 BC128571.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01801700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02161112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01801698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01637951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02161107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01152903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413683 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484819 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484818 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530947 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870368 - -
          Illumina mouse-6 v1.0 expression beadchip 870368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2725727 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2725727 - -
          mouse-6 v1.1 (Illumina) scl4279.1.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 870368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4279.1.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2725727 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2725727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline