Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01339707_g1
          See other XRCC3 GE Assays ›
          Gene Symbol:
          Xrcc3
          Gene Name:
          X-ray repair complementing defective repair in Chinese hamster cells 3
          Gene Aliases:
          4432412E01Rik, AI182522, AW537713
          Chromosome Location:
          Chr.12: 111803192 - 111813841 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01339707_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xrcc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028875.2 NP_083151.1 7 - 8 938 67
          GenBank mRNA AK014491.1 - 7 - 8 938 67
          BC043073.1 - 6 - 7 815 6404521 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002415.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xrcc3

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          regulation of centrosome duplication
          resolution of mitotic recombination intermediates
          positive regulation of mitotic cell cycle spindle assembly checkpoint
          nucleotide binding
          four-way junction DNA binding
          DNA binding
          ATP binding
          DNA-dependent ATPase activity
          crossover junction endodeoxyribonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746253 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746258 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746252 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746257 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152049 BC043073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152049 NM_028875.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152049 AK014491.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135441 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134513 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152049 BC043073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152049 NM_028875.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152049 AK014491.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197043 BC043073.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197043 NM_028875.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197043 AK014491.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28746239 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134513 - -
          Whole Mouse Genome G4122A A_51_P197043 BC043073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197043 NM_028875.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197043 AK014491.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119265 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134513 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09860345 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09860347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10200648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10200647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746252 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746258 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28746253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28746183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10132857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09818697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10132859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10094712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10406678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10095986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10371800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10132860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09818692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10094715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09987897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10200649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402729 - -
          Affymetrix HT MG-430 PM Array Plate 1437623_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1452748_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10402721 - -
          Mouse Genome 430 2.0 Array 1452748_at - -
          Mouse Genome 430 2.0 Array 1437623_x_at - -
          Mouse Genome 430 2.0 Array 1452748_PM_at - -
          Mouse Genome 430 2.0 Array 1437623_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74335_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74335_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200016763.mm.1 NM_028875.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016763.mm.1 TC1200002415.mm.1 MTA TC
          Mouse Expression Set 430 1437623_x_at - -
          Mouse Expression Set 430 1437623_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1437623_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1452748_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452748_at - -
          Affymetrix HT Mouse Genome 430B Array 1437623_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 765900 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760551 - -
          Illumina mouse-6 v1.0 expression beadchip 6760551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2496421 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2496421 - -
          mouse-6 v1.1 (Illumina) scl42028.9_457-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42028.9_457-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2496421 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2496421 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline