Search
Search

Fbxo18| Gene Symbol: Fbxo18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015792.1 | NP_056607.1 | 6 - 7 | 1293 | 117 | |
| GenBank mRNA | AF184275.1 | - | 4 - 5 | 787 | 117 | |
| AK083118.1 | - | 6 - 7 | 1638 | 117 | ||
| AK084645.1 | - | 6 - 7 | 1229 | 117 | ||
| AK147829.1 | - | 6 - 7 | 1277 | 117 | ||
| AK154357.1 | - | 6 - 7 | 1316 | 117 | ||
| AK158937.1 | - | 6 - 7 | 1344 | 117 | ||
| BC031393.1 | - | 6 - 7 | 1293 | 4948804 | 117 | |
Set Membership: |
|