Search
Search
| Gene Symbol: Gcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC110631.1 | - | 2 - 3 | 404 | 40054293 | 72 |
| BC110632.1 | - | 2 - 3 | 404 | 40054295 | 72 | |
| D88611.1 | - | 2 - 3 | 581 | 72 | ||
Set Membership: |
|