Search
Search
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110780.1 | NP_001104250.1 | 11 - 12 | 1526 | 121 | |
| NM_013680.4 | NP_038708.3 | 11 - 12 | 1526 | 121 | ||
| GenBank mRNA | BC022954.1 | - | 11 - 12 | 1504 | 5368098 | 121 |
Set Membership: |
|