Search
Search
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030235.1 | NP_084511.1 | 11 - 12 | 1566 | 64 | |
| GenBank mRNA | AK020003.1 | - | 11 - 12 | 1543 | 64 | |
| AK083605.1 | - | 11 - 12 | 1523 | 64 | ||
| AK122228.1 | - | 11 - 12 | 1544 | 64 | ||
| AK147365.1 | - | 11 - 12 | 1568 | 64 | ||
| BC117878.1 | - | 11 - 12 | 1528 | 40095961 | 64 | |
| BC117879.1 | - | 11 - 12 | 1528 | 64 | ||
Set Membership: |
|