Search
Search

Capzb| Gene Symbol: Capzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037761.2 | NP_001032850.1 | 2 - 3 | 219 | 72 | |
| NM_001271405.1 | NP_001258334.1 | 2 - 3 | 198 | 72 | ||
| NM_001271406.1 | NP_001258335.1 | 2 - 3 | 184 | 72 | ||
| NM_009798.4 | NP_033928.1 | 2 - 3 | 219 | 72 | ||
| GenBank mRNA | AK153367.1 | - | 2 - 3 | 150 | 72 | |
| AK153394.1 | - | 2 - 3 | 150 | 72 | ||
| AK156778.1 | - | 2 - 3 | 188 | 72 | ||
| AK159390.1 | - | 2 - 3 | 193 | 72 | ||
| AK160082.1 | - | 2 - 3 | 194 | 72 | ||
| AK162757.1 | - | 2 - 3 | 186 | 72 | ||
| AK168738.1 | - | 2 - 3 | 147 | 72 | ||
| BC002053.1 | - | 2 - 3 | 161 | 3590980 | 72 | |
| BY124858.1 | - | 2 - 3 | 208 | 72 | ||
| FJ692320.1 | - | 2 - 3 | 184 | 72 | ||
| U10406.1 | - | 2 - 3 | 139 | 72 | ||
| U10407.1 | - | 2 - 3 | 110 | 72 | ||
Set Membership: |
|