Search
Search
| Gene Symbol: Spen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019763.2 | NP_062737.2 | 3 - 4 | 1242 | 98 | |
| GenBank mRNA | AB055980.1 | - | 2 - 3 | 684 | 95 | |
| AF156529.1 | - | 2 - 3 | 744 | 95 | ||
| AK137647.1 | - | 3 - 4 | 1130 | 98 | ||
Set Membership: |
|