| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2115310 |
BC112427.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2115310 |
BC113132.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2115310 |
NM_177867.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2115310 |
AK077155.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955188 |
NM_177867.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955188 |
AK077155.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268107 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
373768 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
869857 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759257 |
AK050955.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759257 |
AK077394.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759257 |
AK161040.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
759257 |
NM_028248.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415420 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702741 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
405184 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
120952 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
81182 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
33571 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
11209 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
488527 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
637577 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
624589 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
151421 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861954 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
919778 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
144976 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65786 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
108667 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P108031 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955188 |
NM_177867.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955188 |
AK077155.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2115310 |
BC112427.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2115310 |
BC113132.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2115310 |
NM_177867.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2115310 |
AK077155.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362056 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362063 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362060 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362057 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362054 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362059 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23362064 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01006857 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01006844 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P108031 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155255 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P210059 |
BC112427.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P210059 |
BC113132.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P210059 |
NM_177867.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P210059 |
AK077155.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131736 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01006844 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P108031 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155255 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03224122 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03224123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362048 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362254 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362270 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362119 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362082 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362217 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362263 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362261 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362088 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362265 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362105 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362029 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362076 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362164 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362253 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362118 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362052 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362097 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362019 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362139 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362259 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362151 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362100 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362190 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362236 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362064 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362145 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362108 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362122 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362174 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362086 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362056 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362248 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362264 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362144 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362128 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362066 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362130 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362239 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23362134 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362056 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362048 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362189 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362190 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03623431 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362179 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04066021 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03835175 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04063969 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362075 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03551166 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362091 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03523906 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03753756 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362054 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362155 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362061 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03986844 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362170 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03441357 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362036 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362058 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362052 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362086 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362156 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03441352 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03623429 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03811824 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362180 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362035 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362051 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362078 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362168 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362183 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362060 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362191 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362159 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362088 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03441356 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03604924 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03961290 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03986845 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03623426 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362080 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04066026 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362084 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362072 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04066020 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03858373 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03933193 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03894226 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362178 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03521503 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362165 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362171 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23362073 |
- |
- |