Search
Search

Ercc4| Gene Symbol: Ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015769.2 | NP_056584.2 | 2 - 3 | 591 | 81 | |
| GenBank mRNA | AF189285.1 | - | 2 - 3 | 591 | 81 | |
| AK018375.1 | - | 2 - 3 | 411 | 81 | ||
| AK138588.1 | - | 2 - 3 | 385 | 81 | ||
| AK149326.1 | - | 2 - 3 | 424 | 81 | ||
| BC026792.1 | - | 2 - 3 | 382 | 4911382 | 81 | |
| BC051036.1 | - | 1 - 2 | 122 | 81 | ||
Set Membership: |
|