Search
Search

Upk1b| Gene Symbol: Upk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178924.4 | NP_849255.2 | 2 - 3 | 137 | 106 | |
| GenBank mRNA | AF274865.1 | - | 1 - 2 | 72 | 106 | |
| AK053004.1 | - | 2 - 3 | 138 | 106 | ||
| AK083101.1 | - | 2 - 3 | 137 | 106 | ||
| AK086926.1 | - | 2 - 3 | 136 | 106 | ||
| BC046604.1 | - | 2 - 3 | 115 | 6389837 | 106 | |
Set Membership: |
|