Search
Search
| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038018.4 | NP_001033107.1 | 8 - 9 | 914 | 94 | |
| NM_001112711.2 | NP_001106182.1 | 8 - 9 | 914 | 94 | ||
| NM_001286063.1 | NP_001272992.1 | 8 - 9 | 894 | 94 | ||
| NM_001286064.1 | NP_001272993.1 | 8 - 9 | 899 | 94 | ||
| NM_011938.4 | NP_036068.2 | 8 - 9 | 914 | 94 | ||
| GenBank mRNA | AF040747.1 | - | 8 - 9 | 784 | 94 | |
| AF040748.1 | - | 8 - 9 | 784 | 94 | ||
| AF040749.1 | - | 8 - 9 | 784 | 94 | ||
| AK051405.1 | - | 8 - 9 | 896 | 94 | ||
| AK161142.1 | - | 6 - 7 | 546 | 94 | ||
| BC057206.1 | - | 8 - 9 | 888 | 94 | ||
| BC075719.1 | - | 8 - 9 | 795 | 5031455 | 94 | |
| Y15797.1 | - | 8 - 9 | 736 | 94 | ||
| Y15798.1 | - | 8 - 9 | 758 | 94 | ||
| Y15799.1 | - | 8 - 9 | 741 | 94 | ||
Set Membership: |
|