Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01342560_m1
          See other SLC6A20B GE Assays ›
          Gene Symbol:
          Slc6a20b
          Gene Name:
          solute carrier family 6 (neurotransmitter transporter), member 20B
          Gene Aliases:
          AA986306, Sit1, Slc6a20, XT3, Xtrp3
          Chromosome Location:
          Chr.9: 123593820 - 123632565 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01342560_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc6a20b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011731.3 NP_035861.2 2 - 3 595 79
          GenBank mRNA AF075261.1 - 2 - 3 419 79
          AK076806.1 - 3 - 4 329 79
          AK143893.1 - 2 - 3 565 79
          AK144038.1 - 2 - 3 596 79
          AK163340.1 - 3 - 4 484 79
          AK164706.1 - 2 - 3 440 79
          AK164713.1 - 2 - 3 430 79
          BC013484.1 - 2 - 3 506 4242168 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003306.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc6a20b

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          neurotransmitter transport
          transporter activity
          neurotransmitter:sodium symporter activity
          protein binding
          symporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900027867.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027867.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027867.mm.1 TC0900003306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027870.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027870.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027870.mm.1 TC0900003306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027866.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027866.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027866.mm.1 TC0900003306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027868.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027868.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027868.mm.1 TC0900003306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027865.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027865.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027865.mm.1 TC0900003306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027864.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027864.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027864.mm.1 TC0900003306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027869.mm.1 NM_011731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027869.mm.1 NM_011731.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027869.mm.1 TC0900003306.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline