Search
Search

Prkdc| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011159.2 | NP_035289.2 | 4 - 5 | 416 | 149 | |
| GenBank mRNA | AB007544.1 | - | 4 - 5 | 393 | 149 | |
| AB011543.1 | - | 4 - 5 | 405 | 149 | ||
| AK084827.1 | - | 4 - 5 | 417 | 149 | ||
| D87521.1 | - | 4 - 5 | 393 | 149 | ||
Set Membership: |
|