Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01343295_gH
          See other 4930453N24RIK GE Assays ›
          Gene Symbol:
          4930453N24Rik
          Gene Name:
          RIKEN cDNA 4930453N24 gene
          Gene Aliases:
          AI649104
          Chromosome Location:
          Chr.16: 64762152 - 64771156 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01343295_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930453N24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026273.2 NP_080549.1 2 - 3 443 87
          GenBank mRNA AK002578.1 - 2 - 3 444 87
          AK161311.1 - 2 - 3 444 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821648 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129672 AK075826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129672 AK161311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129672 BC020029.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129672 NM_026273.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57346 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127577 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166286 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124902 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129672 AK075826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129672 AK161311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129672 BC020029.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129672 NM_026273.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127577 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124902 - -
          Whole Mouse Genome G4122A A_52_P351311 AK161311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P351311 BC020029.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P351311 NM_026273.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P351311 AK002578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311621 AK075826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311621 AK002578.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129672 AK075826.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129672 AK161311.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129672 BC020029.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129672 NM_026273.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127577 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124902 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12995485 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12995486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12953013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12598750 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821648 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30821643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30821642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12953013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12995486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12915510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12510932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12490210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12594334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12569246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12915516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12759528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12490198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12666021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13049159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30821627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440287 - -
          Affymetrix HT MG-430 PM Array Plate 1449719_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423976_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449718_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1438553_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10440284 - -
          Mouse Genome 430 2.0 Array 1438553_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438553_x_at - -
          Mouse Genome 430 2.0 Array 1449719_PM_at - -
          Mouse Genome 430 2.0 Array 1449719_at - -
          Mouse Genome 430 2.0 Array 1423977_at - -
          Mouse Genome 430 2.0 Array 1423976_PM_at - -
          Mouse Genome 430 2.0 Array 1449718_s_at - -
          Mouse Genome 430 2.0 Array 1427664_PM_at - -
          Mouse Genome 430 2.0 Array 1427664_at - -
          Mouse Genome 430 2.0 Array 1449718_PM_s_at - -
          Mouse Genome 430 2.0 Array 1423976_at - -
          Mouse Genome 430 2.0 Array 1423977_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67609_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67609_at - -
          Affymetrix Mouse Clariom S Array PSR1600014684.mm.1 NM_026273.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014684.mm.1 NM_026273.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014684.mm.1 TC1600001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600014685.mm.1 NM_026273.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014685.mm.1 NM_026273.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014685.mm.1 TC1600001837.mm.1 MTA TC
          Mouse Expression Set 430 1423977_PM_at - -
          Mouse Expression Set 430 1423976_at - -
          Mouse Expression Set 430 1423976_PM_at - -
          Mouse Expression Set 430 1423977_at - -
          Murine Genome U74v2 Set 94861_PM_at - -
          Murine Genome U74v2 Set 94861_at - -
          Murine Genome U74v2 Set 169337_at - -
          Murine Genome U74v2 Set 169337_PM_at - -
          Mouse Genome 430A 2.0 Array 1423976_at - -
          Mouse Genome 430A 2.0 Array 1423976_PM_at - -
          Mouse Genome 430A 2.0 Array 1423977_at - -
          Mouse Genome 430A 2.0 Array 1423977_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449718_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1423976_at - -
          Affymetrix HT Mouse Genome 430A Array 1449719_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423976_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449719_at - -
          Affymetrix HT Mouse Genome 430A Array 1449718_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 519746 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103710053 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220556 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2888766 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2888766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline