Search
Search

Scaf4| Gene Symbol: Scaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178923.3 | NP_849254.2 | 7 - 8 | 1137 | 69 | |
| GenBank mRNA | AK045266.1 | - | 7 - 8 | 1134 | 69 | |
| AK173120.1 | - | 7 - 8 | 1215 | 69 | ||
| BC023734.1 | - | 4 - 5 | 538 | 69 | ||
| BC041782.1 | - | 4 - 5 | 509 | 69 | ||
| BC053096.1 | - | 7 - 8 | 1032 | 6401143 | 69 | |
| BC057592.1 | - | 7 - 8 | 1137 | 6417362 | 69 | |
Set Membership: |
|