Search
Search

Psmd2| Gene Symbol: Psmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134101.2 | NP_598862.1 | 5 - 6 | 772 | 65 | |
| NR_027485.1 | 5 - 6 | 864 | 65 | |||
| GenBank mRNA | AK146482.1 | - | 5 - 6 | 721 | 65 | |
| AK150374.1 | - | 5 - 6 | 744 | 65 | ||
| AK151588.1 | - | 5 - 6 | 744 | 65 | ||
| AK152007.1 | - | 5 - 6 | 744 | 65 | ||
| AK152970.1 | - | 5 - 6 | 742 | 65 | ||
| AK159098.1 | - | 5 - 6 | 724 | 65 | ||
| AK159636.1 | - | 5 - 6 | 721 | 65 | ||
| AK159756.1 | - | 5 - 6 | 744 | 65 | ||
| AK159815.1 | - | 5 - 6 | 740 | 65 | ||
| AK166816.1 | - | 5 - 6 | 724 | 65 | ||
| AK167326.1 | - | 5 - 6 | 724 | 65 | ||
| AK167774.1 | - | 5 - 6 | 721 | 65 | ||
| AK167786.1 | - | 5 - 6 | 722 | 65 | ||
| AK167909.1 | - | 5 - 6 | 732 | 65 | ||
| AK167995.1 | - | 5 - 6 | 740 | 65 | ||
| AK168131.1 | - | 5 - 6 | 731 | 65 | ||
| AK168398.1 | - | 5 - 6 | 724 | 65 | ||
| AK168543.1 | - | 5 - 6 | 721 | 65 | ||
| AK169867.1 | - | 5 - 6 | 724 | 65 | ||
| AK202594.1 | - | 1 - 2 | 167 | 65 | ||
| BC021508.1 | - | 5 - 6 | 741 | 5347139 | 65 | |
| BC024058.1 | - | 5 - 6 | 714 | 65 | ||
| BC024830.1 | - | 5 - 6 | 712 | 5358424 | 65 | |
| BC027388.1 | - | 5 - 6 | 736 | 4952935 | 65 | |
| BC027767.1 | - | 5 - 6 | 740 | 5347281 | 65 | |
| BC027822.1 | - | 5 - 6 | 723 | 5321505 | 65 | |
| BC028851.1 | - | 5 - 6 | 712 | 3660169 | 65 | |
| BC031128.1 | - | 5 - 6 | 741 | 4982313 | 65 | |
| BC033325.1 | - | 5 - 6 | 801 | 65 | ||
Set Membership: |
|