Search
Search

Myh11| Gene Symbol: Myh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161775.1 | NP_001155247.1 | 33 - 34 | 4944 | 63 | |
| NM_013607.2 | NP_038635.2 | 33 - 34 | 4944 | 63 | ||
| GenBank mRNA | AK173045.1 | - | 33 - 34 | 4908 | 63 | |
| BC026142.1 | - | 33 - 34 | 4929 | 3989495 | 63 | |
| D85923.1 | - | 33 - 34 | 4899 | 63 | ||
| D85924.1 | - | 33 - 34 | 4899 | 63 | ||
Set Membership: |
|