Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344436_g1
          See other PLA2G10 GE Assays ›
          Gene Symbol:
          Pla2g10
          Gene Name:
          phospholipase A2, group X
          Gene Aliases:
          PLA2GX, mGXsPLA2, sPLA2-X
          Chromosome Location:
          Chr.16: 13715057 - 13730983 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344436_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291009.1 NP_001277938.1 2 - 3 779 84
          NM_011987.3 NP_036117.1 2 - 3 779 84
          NR_110990.1 2 - 3 779 84
          GenBank mRNA AF166097.2 - 2 - 3 381 84
          AK076921.1 - 2 - 3 179 84
          BC028879.1 - 2 - 3 239 4218273 84
          KC789644.1 - 2 - 3 225 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001233.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pla2g10

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phospholipid metabolic process
          axon guidance
          positive regulation of lipid storage
          lipid catabolic process
          positive regulation of cellular protein metabolic process
          positive regulation of prostaglandin secretion
          cholesterol homeostasis
          regulation of macrophage activation
          negative regulation of sequence-specific DNA binding transcription factor activity
          lysophospholipid transport
          positive regulation of arachidonic acid secretion
          negative regulation of cholesterol efflux
          phospholipase activity
          phospholipase A2 activity
          calcium ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035772 AK076921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035772 BC028879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035772 NM_011987.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035772 AF166097.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903511 XM_001473978.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903511 XM_980722.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158405 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104083 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169903 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035772 AK076921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035772 BC028879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035772 NM_011987.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035772 AF166097.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P380451 AK076921.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P380451 BC028879.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P380451 NM_011987.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P380451 AF166097.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30558749 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104083 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08513560 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169903 - -
          Whole Mouse Genome G4122A A_51_P380451 AK076921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P380451 BC028879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P380451 NM_011987.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P380451 AF166097.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102031 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104083 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12617747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12829026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12617750 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12617748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12617753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12617749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12829016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30558714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12977241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12574908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12704391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12892226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12724329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12925928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12704394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12925926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12849108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13042844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12900048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12588003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12704392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30558717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10437810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437805 - -
          Affymetrix HT MG-430 PM Array Plate 1451502_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10437804 - -
          Mouse Genome 430 2.0 Array 1451502_PM_at - -
          Mouse Genome 430 2.0 Array 1451502_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26565_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26565_at - -
          Affymetrix Mouse Clariom S Array PSR1600009859.mm.1 NM_011987.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009859.mm.1 TC1600001233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009858.mm.1 TC1600001233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009860.mm.1 NM_011987.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009860.mm.1 TC1600001233.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1451502_at - -
          Affymetrix HT Mouse Genome 430A Array 1451502_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609753 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609753 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline