Search
Search

Thap7| Gene Symbol: Thap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026909.2 | NP_081185.1 | 3 - 4 | 266 | 102 | |
| GenBank mRNA | AK007329.1 | - | 2 - 3 | 218 | 102 | |
| AK154641.1 | - | 3 - 4 | 352 | 102 | ||
| BC018221.1 | - | 3 - 4 | 266 | 4215606 | 102 | |
Set Membership: |
|