Search
Search

Ppef2| Gene Symbol: Ppef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011148.3 | NP_035278.1 | 14 - 15 | 1928 | 70 | |
| GenBank mRNA | AF023458.1 | - | 14 - 15 | 1928 | 70 | |
| BC027049.1 | - | 15 - 16 | 2235 | 5356685 | 70 | |
Set Membership: |
|