Search
Search

Zcchc7| Gene Symbol: Zcchc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138590.4 | NP_613056.2 | 7 - 8 | 1220 | 95 | |
| GenBank mRNA | AK013175.1 | - | 1 - 2 | 704 | 95 | |
| AK050430.1 | - | 1 - 2 | 1912 | 95 | ||
| BC050200.1 | - | 6 - 7 | 95 | |||
| BC054405.1 | - | 1 - 2 | 423 | 95 | ||
Set Membership: |
|